Learn More
Abeba™ Protektor Line 2232 Shoes
Product Code. 15608037
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15608037
Brand: Abeba™ 2232/36
Only null left
Add to basket
Add to basket
Leather upper construction includes reflective material.
  • CE EN ISO 20345:2011 S3 SRC
  • Smooth leather
  • Black
  • Steel toe cap
  • Penetration resistant steel midsole
  • Slip resistance PU outsole sole
  • Antistatic
  • Leather lining
  • Double velcro fastening
Specifications
Certifications/Compliance CE EN ISO 20345:2011 S3 SRC
Color Black
Material Leather
Size 36
Sole Features Slip-resistant
Sole Material Polyurethane
Style 2232
Toe Material Steel