Learn More
Protein kinase inhibitors 1 hydrochloride, MedChemExpress
Product Code. 16495437
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495437 50 mg 50mg
16459306 100 mg 100mg
16454106 25 mg 25mg
16442648 5 mg 5mg
16430449 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16495437 Supplier Medchem Express Supplier No. HYU00439A
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
MedChemExpress Protein kinase inhibitors 1 hydrochloride is a potent HIPK2 inhibitor, with IC50s of 136 and 74 nM for HIPK1 and HIPK2, and a Kd of 9.5 nM for HIPK2.
Protein kinase inhibitors 1 hydrochloride is a potent HIPK2 inhibitor, with IC50s of 136 and 74 nM for HIPK1 and HIPK2, and a Kd of 9.5 nM for HIPK2.
Specifications
Chemical Name or Material Protein kinase inhibitors 1 hydrochloride
CAS 2321337-71-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H18ClN5O3S
Quantity 50 mg
Solubility Information H2O : 9.09 mg/mL (21.65 mM; Need ultrasonic) ∣DMSO : 8.33 mg/mL (19.84 mM; Need ultrasonic)
SMILES O=C(NC/1=O)SC1=C\C2=CNC(C(C3=CC=C(N4CCNCC4)N=C3)=C2)=O.[H]Cl
Molecular Weight (g/mol) 419.89
Formula Weight 419.89
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease