missing translation for 'onlineSavingsMsg'
Learn More

Protein kinase inhibitors 1 hydrochloride, MedChemExpress

Product Code. 16454106
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16454106 25 mg 25mg
16495437 50 mg 50mg
16459306 100 mg 100mg
16442648 5 mg 5mg
16430449 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16454106 Supplier Medchem Express Supplier No. HYU00439A
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Protein kinase inhibitors 1 hydrochloride is a potent HIPK2 inhibitor, with IC50s of 136 and 74 nM for HIPK1 and HIPK2, and a Kd of 9.5 nM for HIPK2.

Protein kinase inhibitors 1 hydrochloride is a potent HIPK2 inhibitor, with IC50s of 136 and 74 nM for HIPK1 and HIPK2, and a Kd of 9.5 nM for HIPK2.

Specifications

Chemical Name or Material Protein kinase inhibitors 1 hydrochloride
CAS 2321337-71-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H18ClN5O3S
Quantity 25 mg
Solubility Information H2O : 9.09 mg/mL (21.65 mM; Need ultrasonic) ∣DMSO : 8.33 mg/mL (19.84 mM; Need ultrasonic)
SMILES O=C(NC/1=O)SC1=C\C2=CNC(C(C3=CC=C(N4CCNCC4)N=C3)=C2)=O.[H]Cl
Molecular Weight (g/mol) 419.89
Formula Weight 419.89
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.