Learn More
Thermo Scientific™ Protein Kinase C-epsilon Peptide Substrate
Click to view available options
Substrate Quantity:
0.5mg
1mg
Unit Size:
0.5mg
1mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Color | White |
| Form | Lyophilized |
| Molecular Weight (g/mol) | 2067.45 |
| Storage Requirements | -30° to -10°C |
| Substrate Quantity | 1mg |
For Research Use Only