Learn More
Thermo Scientific™ Protein Kinase C (19-36)
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Purification Method | HPLC |
| Common Name | PKC Inhibitor Peptide |
| Content And Storage | -30°C to -10°C |
| Format | Solid |
| Molecular Weight (g/mol) | 2151.51 |
| Product Type | Protein Kinase C (19-36) |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Sequence | Arg-Phe-Ala-Arg-Lys-Gly-Ala-Leu-Arg-Gln-Lys-Asn-Val-His-Glu-Val-Lys-Asn |
For Research Use Only