Learn More
Corning™ Protein Crystallization Microplates
Product Code. 11339464 Shop All Corning Products
Change view
Click to view available options
Well Shape:
Conical Bottom (V-well)
Unit Size:
Pack of 50
Product Code. Well Shape unitSize
11339464 Conical Bottom (V-well) Pack of 50
1 options
Product Code. 11339464
Brand: Corning™ 3551
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
More hydrophilic to promote crystal centering and reduce wicking
  • For high throughput sitting-drop protein crystallisation
  • Low water absorption of the polymer prevents loss of protein drop volume
  • Conical flat well shape
  • Hydrophilic surface treated
  • COC material
  • Non-sterile
  • Embossed reservoir numbers for easy identification
  • Without lid
  • Allows protein crystals to be easily viewed under polarised light with minimal backgroud interferance
  • High clarity protein wells offer optimal microscopic crystal viewing
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Lid Without Lid
Description Made from Cyclic Olefin Copolymer (COC), Conical flat
Array 8 x 12
Color Clear
No. of Wells 96
Sterility Non-sterile
Surface Treatment Hydrophilic
Well Shape Conical Bottom (V-well)
For Use With (Application) Protein crystallisation
Profile Conical Flat
Quantity 50/Cs.