Learn More
Labconco™ Protector™ FilterMate™ Filter Cells and Prefilters
High-efficiency filter cells for use with Labconco™ FilterMate™ Portable Exhausters
15610.00 NOK - 16405.00 NOK
Specifications
For Use With (Equipment) FilterMate™ Portable Exhauster
View More Specs
Products 2
Product Code Brand Weight (English) Filter Type Product Type Weight (Metric) Price Quantity & Availability  
Product Code Brand Weight (English) Filter Type Product Type Weight (Metric) Price Quantity & Availability  
16283251
View Documents Promotion Details
Labconco™
6910401
45 lb. Carbon Organic (AC) Filter Cell 20.4 Kg
16405.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 13-08-2024
to see stock.
16293251
View Documents Promotion Details
Labconco™
6933201
45 lb. HEPA HEPA Filter Cell 20.4 Kg
15610.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 13-08-2024
to see stock.
Description
Description
Four carbon-based ColorSmart filter cells with a minimum of 26 lb. (12kg) of impregnated carbon are available to adsorb or treat chemical fumes and vapors from ammonia, acid gases, organics and formaldehyde.
  • Each ammonia, acid gases and formaldehyde filter cell comes with three contaminant detector tubes
  • For particulates such as powders and pollens, a ColorSmart HEPA filter cell is available to efficiently remove 99.99% of aerosols and particulate matter 0.3 micron in diameter or larger
  • Filter cells are engineered to NIOSH guidelines for respirators
  • Filter cells are color coded the same as respirator cartridge filter types for easy, at-a-glance recognition
  • Roughing prefilter removes dust and particulates to extend the life of filter cells
Order Info
Optional electrostatic prefilter removes mists and aerosols. (Prefilters may be omitted when using HEPA filter cell.)
Specifications
Specifications
FilterMate™ Portable Exhauster