Learn More
Mutexil™ Protective Overshoe Coverings
Description
- Various sizes and colors
- Needs-based styles
- Per new standards EN ISO 20345 and EN ISO 20347
Specifications
Specifications
| Color | Blue |
| For Use With (Application) | For sanitary, food, industrial, catering and services |
| Material | Polyethylene |
| Size | Universal |
| Style | CPE |
| Waterproof | Yes |
| Product Type | Protective CPE Visitor Overshoes |