Learn More
Mutexil™ Protective Overshoe Coverings
Product Code. 11770124
Change view
Click to view available options
Product Type:
Overshoe
Protective CPE Visitor Overshoes
Sticky Overshoe
Color:
Blue
White
Size:
2X-Large
Universal
X-Large
Unit Size:
Pack of 2000
Pack of 300
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Color Size Unit Size
11770124 Protective CPE Visitor Overshoes Blue X-Large Pack of 2000
11780124 Protective CPE Visitor Overshoes Blue Universal Pack of 2000
11790124 Protective CPE Visitor Overshoes White Universal Pack of 2000
11775433 Sticky Overshoe White 2X-Large Pack of 300
11765433 Overshoe Blue 2X-Large Pack of 300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11770124 Supplier Mutexil™ Supplier No. 2000019
  • ONLINE EXCLUSIVE PRICE
    1001.19 NOK
    558.30 NOK / Pack of 2000
    Save 442.89 NOK
    44% Off
Estimated Shipment: 16-10-2026
Choose from a range of choices and construction, from sticky, anti-slip, fluid -resistant, polypropylene and CPE variations with Mutexil™ Protective Overshoe Coverings.
  • Various sizes and colors
  • Needs-based styles
  • Per new standards EN ISO 20345 and EN ISO 20347
Specifications
Color Blue
For Use With (Application) For sanitary, food, industrial, catering and services
Material Polyethylene
Size X-Large
Style CPE
Waterproof Yes
Product Type Protective CPE Visitor Overshoes