missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteasome 19S S3 Antibody (OTI2A2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-01808
This item is not returnable.
View return policy
Description
Proteasome 19S S3 Monoclonal specifically detects Proteasome 19S S3 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| Proteasome 19S S3 | |
| Monoclonal | |
| 0.95 mg/mL | |
| Western Blot 1:500-2000, Flow Cytometry 1:100 | |
| PSMD3 | |
| Full length human recombinant protein of human PSMD3(NP_002800) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG2a |
| Western Blot, Flow Cytometry | |
| OTI2A2 | |
| Unconjugated | |
| 26S proteasome regulatory subunit RPN3, 26S proteasome regulatory subunit S3, P58, proteasome (prosome, macropain) 26S subunit, non-ATPase, 3, Proteasome subunit p58, RPN3, S3, 26S proteasome non-ATPase regulatory subunit 3, tissue specific transplantation antigen 2, TSTA2 | |
| Mouse | |
| 60.8 kDa | |
| 0.1 mL | |
| Neuroscience, Ubiquitin Proteasome Pathway | |
| 5709 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction