Learn More
Contec™ PROSAT™ Sterile™ Meltblown Polypropylene Wipes
Shop All Contec Products
Description
Wipe down isolators, cabinets, hoods, and other cleanroom equipment and tools. Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes are ideal for cleanup following BD RODAC™ plate sampling. These wipes are designed specifically for steril
These PROSAT™ presaturated wipes are exceptionally clean and also free from additives of any kind. This particular meltblown polypropylene contains very low levels of sodium and other ions. PROSAT™ meltblown polypropylene wipes are ideal for wiping articles prior to pass through, cleaning process tools and other equipment, and general surface cleaning.
- Low in particles and fibers, making it ideal for product contact areas
- Presaturated wipes reduce solvent usage and VOC emissions and ensure consistent saturation of each wipe independent of the operator
- Validated sterile via gamma irradiation to a 10-6 Sterility Assurance Level
- Compatible with Grade A/B or ISO Class 5 through 8 environments
- Wipes are provided in a resealable pouch for ease of use and convenience
Specifications
Specifications
| Product Type | Pre-saturated Wipe |
| Length (English) | 11 in. |
| Length (Metric) | 28 cm |
| Width (English) | 9 in. |
| Width (Metric) | 23 cm |
| Material | Polypropylene |
| Certifications/Compliance | ISO Class 5 to 8 |
| Dimensions (L x W) | 28 x 23 cm (11 x 9 in.) |
| For Use With (Application) | Routine Equipment Cleaning in Sterile Environments |
| Packaging Quantity | 30 Wipes/Pk., 48 Pk./Cs. |
| Show More |
Safety and Handling
- CONTEC PROSAT MBPP WITH 70% IPA
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P370+P378-In case of fire: Use dry chemical, carbondioxide to extinguish.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : PROPAN-2-OL