Learn More
Propionic acid, Sigma-Aldrich™
Specifications
Specifications
| Chemical Name or Material | Propionic acid |
| CAS | 79-09-4 |
| Boiling Range | 140.7°C to 141.4°C/1013 hPa |
| Melting Point | -20°C |
| Density | 0.993 g/cm³ at 20°C |
| Flash Point | 54°C (closed cup) |
| Quantity | 25 L |
| Synonym | Propionic acid, Methyl acetic acid, Propionic acid |
| Molecular Weight (g/mol) | 74.08 |
| Grade | Not Available |