Learn More
Propionic Acid Hemicalcium Salt, MP Biomedicals™
Product Code. 11471802 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471802 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11471802 Supplier MP Biomedicals Supplier No. 0210498791
  • ONLINE EXCLUSIVE PRICE
    6150.00 NOK
    5858.17 NOK / 1kg
    Save 291.83 NOK
    4% Off
 Restricted Item
Estimated Shipment: 04-02-2027
An antifungal agent. Soluble in water (1g/3mL); slightly soluble in methanol, ethanol; practically insoluble in acetone, benzene
Specifications
CAS 4075-81-4
Color White
Molecular Formula C6H10CaO4
MDL Number MFCD00167354
Quantity 1 kg
Synonym calcium propionate, calcium dipropionate, calcium propanoate, mycoban, propanoic acid, calcium salt, bioban-c, caswell no. 151, propionic acid, calcium salt, unii-8ai80040kw, hsdb 907
InChI Key BCZXFFBUYPCTSJ-UHFFFAOYSA-L
SMILES [Ca++].CCC([O-])=O.CCC([O-])=O
IUPAC Name calcium dipropanoate
Molecular Weight (g/mol) 186.22
PubChem CID 19999
ChEBI CHEBI:81716
Formula Weight 93.1
Physical Form Powder
Chemical Name or Material Propionic Acid Hemicalcium Salt