Learn More
Propionaldehyde, Sigma-Aldrich™
Product Code. 15861681
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15861681 1 L 1 L
15851681 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15861681 Supplier Sigma Aldrich Supplier No. 8221331000
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 123-38-6
Specifications
Density 0.81 g/cm3 at 20°C
Boiling Point 47°C to 48°C/1013 hPa
Quantity 1 L
Grade Research