Learn More
Properidine (1-Methylethyl 1-Methyl-4-phenyl-piperidine-4-carboxylate) 1.0 mg/ml in Methanol, LoGiCal
Product Code. 30457679
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457679 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30457679 Supplier LoGiCal Supplier No. LGCAMP0315.0701
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Properidine (1-Methylethyl 1-Methyl-4-phenyl-piperidine-4-carboxylate)
CAS 561-76-2
Analyte or Component Names Properidine (1-Methylethyl 1-Methyl-4-phenyl-piperidine-4-carboxylate)
Shipping Condition Ice pack (-20°C)
Molecular Formula C16 H23 N O2
Certifications/Compliance ISO 17034
Synonym 1-Methylethyl 1-Methyl-4-phenylpiperidine-4-carboxylate (Properidine), Pethidine Hydrochloride Imp. G (EP), Pethidine Imp. G (EP), Properidine, 1-Methylethyl 1-Methyl-4-phenyl-piperidine-4-carboxylate, 1-Methyl-4-phenyl-4-piperidinecarboxylic acid 1-methylethyl ester, 1-Methyl-4-phenyl-isonipecotic acid isopropyl ester, 1-Methyl-4-phenylpiperidine-4-carboxylic acid isopropyl ester, Gevelina, Ipropethidine, Isopedine, Isopropyl 1-methyl-4-phenylisonipecotate, Isopropyl 1-methyl-4-phenylpiperidine-4-carboxylate, Properidin, Pethidine Hydrochloride Impurity G, Pethidine Impurity G
InChI Formula InChI=1S/C16H23NO2/c1-13(2)19-15(18)16(9-11-17(3)12-10-16)14-7-5-4-6-8-14/h4-8,13H,9-12H2,1-3H3
Solution Type Single Solution
SMILES CC(C)OC(=O)C1(CCN(C)CC1)c2ccccc2
IUPAC Name propan-2-yl 1-methyl-4-phenylpiperidine-4-carboxylate
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 261.36
Quantity 1 mL
Formula Weight 261.1729 g/mol
Show More Show Less
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.