Learn More
Propargyl Sulfide, TRC
Specifica
Specifica
| Chemical Name or Material | Propargyl Sulfide |
| CAS | 13702-09-5 |
| Molecular Formula | C6H6S |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C6H6S/c1-3-5-7-6-4-2/h1-2H,5-6H2 |
| SMILES | C#CCSCC#C |
| IUPAC Name | 3-prop-2-ynylsulfanylprop-1-yne |
| Molecular Weight (g/mol) | 110.18 |
| Formula Weight | 110.019 |