Learn More
Propargyl methacrylate, 98%, stab. with 200-300ppm 4-methoxyphenol, Thermo Scientific Chemicals
Catalog Number 11382487
Click to view available options
Quantity:
10 g
50 g
13861-22-8
C7H8O2
124.14
MFCD00048123
PZAWASVJOPLHCJ-UHFFFAOYSA-N
propargyl methacrylate, 2-propynyl methacrylate, 2-propenoic acid, 2-methyl-, 2-propynyl ester, 2-propenoic acid, 2-methyl-, 2-propyn-1-yl ester, prop-2-yn-1-yl 2-methylprop-2-enoate, prop-2-ynyl methacrylate, propargylmethacrylate, acmc-1bvgj, prop-2-yn-1-yl methacrylate, propargyl methacrylate, stabilized 10g
83778
prop-2-ynyl 2-methylprop-2-enoate
CC(=C)C(=O)OCC#C
13861-22-8
C7H8O2
124.14
MFCD00048123
PZAWASVJOPLHCJ-UHFFFAOYSA-N
propargyl methacrylate, 2-propynyl methacrylate, 2-propenoic acid, 2-methyl-, 2-propynyl ester, 2-propenoic acid, 2-methyl-, 2-propyn-1-yl ester, prop-2-yn-1-yl 2-methylprop-2-enoate, prop-2-ynyl methacrylate, propargylmethacrylate, acmc-1bvgj, prop-2-yn-1-yl methacrylate, propargyl methacrylate, stabilized 10g
83778
prop-2-ynyl 2-methylprop-2-enoate
CC(=C)C(=O)OCC#C
Propargyl methacrylate is a monomer used in polymerization reactions. It is associated with its acetylene and under goes polymerization reaction through the reversible addition fragmentation chain transfer (RAFT) process using cyanoisopropyl dithiobenzoate as a reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Propargyl methacrylate is a monomer used in polymerization reactions. It is associated with its acetylene and under goes polymerization reaction through the reversible addition fragmentation chain transfer (RAFT) process using cyanoisopropyl dithiobenzoate as a reagent.
Solubility
Immiscible with water.
Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 0.986
Boiling Point 149°C to 151°C
Flash Point 48°C (118°F)
Refractive Index 1.45
Quantity 10 g
UN Number UN3272
Beilstein 906850
Solubility Information Immiscible with water.
Formula Weight 124.14
Percent Purity 98%
Chemical Name or Material Propargyl methacrylate, stabilized with 250ppm 4-methoxyphenol