Learn More
Thermo Scientific™ ProPac™ WAX-10 Tertiary Amine Ion Exchange HPLC Column, 10 μm, 22 mm x 250 mm
Column Format: Semi-Preparative Column; Pore Size: Nonporous; Column Phase: Ion Exchange
Brand:  Thermo Scientific™ SP5598
Product Code. 11302493
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
For exceptionally high resolution and efficiency anion-exchange separation of protein variants, use Thermo Scientific™ ProPac™ WAX-10 LC Columns. Based a nonporous 10µm core particle that provides pressure stability, ProPac columns can resolve isoforms that differ by a single charged residue. A thin, hydrophilic layer prevents unwanted secondary interactions. The grafted anion-exchange surface provides pH-based selectivity control and fast mass transfer for high-efficiency separation and moderate capacity.
Maximizing Resolution
  • ProPac WAX-10 LC column is a weak anion-exchanger with a tertiary amine functional group.
  • Column sizes range from 2mm I.D., for analytical work, up to 22mm I.D. for semiprep work.
Superior Performance for protein analysis
  • Ideal for characterization and quality control assessment of closely-related protein variants.
  • Unequaled high-resolution separations.
  • High-efficiency peaks with excellent recovery.
  • Higher capacity than other columns based on nonporous particles.
  • Superior column-to-column and lot-to-lot reproducibility eliminates column variability as a concern in stability and QA/QC testing.