missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Progesterone Recombinant Rabbit Monoclonal Antibody (RM434)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538675
This item is not returnable.
View return policy
Description
Progesterone Recombinant Monoclonal Antibody for ELISA
Progesterone is the major progestational steroid, secreted primarily by the corpus luteum and the placenta. Progesterone binds to the progesterone receptor, resulting in dissociation of heat shock proteins, receptor phosphorylation, and transcription activation through direct or indirect interaction with transcription factors. Progesterone acts on the uterus, the mammary glands and the brain. It is required in embryo implantation, pregnancy maintenance, and the development of mammary tissue for milk production. Progesterone, converted from pregnenolone, also serves as an intermediate in the biosynthesis of gonadal steroid hormones and adrenal corticosteroids. Therapeutic progesterone is a synthetic form of the endogenous hormone progesterone. This agent exerts inhibitory effects on estrogens by decreasing the number of estrogen receptors and increasing its metabolism to inactive metabolites. [National Center for Biotechnology Information. PubChem Compound Database; CID=5994].
Specifications
| Progesterone | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
| Rabbit | |
| Protein A | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
| ELISA | |
| RM434 | |
| Unconjugated | |
| 11-alpha-hydroxyprogesterone; Agolutin; C21H30O2; Crinone; Luteohormone; OHP; Pregn-4-ene-3; Progesterone-11a-hemisuccinate | |
| Progesterone-KLH. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction