Learn More
Productainer™ 3D BioProcess Container, BioTitan retainers, three ports, MPXs
GREENER_CHOICE
Product Code. 17928241 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
100 L
500 L
2000 L
Unit Size:
Pack of 2
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
17928241 100 L Pack of 5
17908281 2000 L Pack of 5
17938281 500 L Pack of 5
31940837 500 L Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17928241 Supplier Thermo Scientific Supplier No. SH31304.01
  • ONLINE EXCLUSIVE PRICE
    98495.00 NOK
    83520.98 NOK / Pack of 5
    Save 14974.02 NOK
    15% Off
 Restricted Item
Estimated Shipment: 10-09-2026
Specifications
Green Features 200 L
Capacity (Metric) 100 L
Film Description 100 L
For Use With (Equipment) 50 L
For Use With (Application) Single-Use Bioprocessing
Quantity 200 L
Product Line 50 L
Tubing Style C-Flex