Learn More
Thermo Scientific™ Productainer™ 3D BioProcess Container, BioTitan retainers, four ports, MPX and Luer locks
Shop All Thermo Scientific Products
Description
Productainer BPCs are constructed with our Thermo Scientific Aegis5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility. Our standard Productainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Productainers can be customized using the industry's largest component library.
Features include:
- Film optimized for flexibility and durability: superior stress resistance, puncture resistance and physical barrier properties to ensure stability of contents
- Gamma-irradiation sterilized
- Squared tube design with polyethylene ports located on the top of the BPC for easy access
- Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
- Customizable via the industry's largest library of qualified components
- Now manufactured with the BioTitan Retention Device—designed to provide superior universal tubing retention that minimizes the risk of leakage and failure at connection points
Recommended for:
- Harvest—collection and storage of bulk product from bioreactors
- Storage—sterile storage of media, buffers and process intermediates
- Separation—feeding and receiving of sterile liquids from separation systems
- Purification—buffer feeding to purification system, fraction collection and storage
- Filling—sterile reservoir for filling systems
- Transport and Shipping—bulk sterile liquids, buffers, media and process intermediates within and between facilities
Specifications
Specifications
| Capacity (Metric) | 50 L |
| For Use With (Application) | Single-Use Bioprocessing |
| Quantity | 50 L |
| Tubing Style | TPE |