Learn More
Velp Scientifica™ Product Manual
Product Code. 15319624 Shop All Velp Products
Click to view available options
For Use With (Equipment):
COEX
CSF6
FIWE Series Raw Fiber Extractors
GDE
HU6
JP
NDA 701 and 702
OXITEST
SER 148 Series Solvent Extractors
SMS
Unit Size:
Each
Product Code. 15319624
Brand: Velp Scientifica™ A00000251
Only null left
Add to basket
Add to basket
Velp Scientifica™ Product Manual are for use with a wide range of products.
  • Inlcudes one manual for the desiered product
  • For use with a wide range of Velp Scientifica™ products
Specifications
Format Manual
For Use With (Equipment) HU6
Product Type Operating Instructions
Quantity 1/Ea.