Description
- Inlcudes one manual for the desiered product
- For use with a wide range of Velp Scientifica™ products
Specifications
Specifications
| Format | Manual |
| For Use With (Equipment) | COEX |
| Product Type | Operating Instructions |
| Quantity | 1/Ea. |