Learn More
Velp Scientifica™ Product Manual
Shop All Velp Products
Click to view available options
For Use With (Equipment):
COEX
CSF6
FIWE Series Raw Fiber Extractors
GDE
HU6
JP
NDA 701 and 702
OXITEST
SER 148 Series Solvent Extractors
SMS
Unit Size:
Each
Description
- Inlcudes one manual for the desiered product
- For use with a wide range of Velp Scientifica™ products
Specifications
Specifications
| Format | Manual |
| For Use With (Equipment) | NDA 701 and 702 |
| Product Type | Operating Instructions |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur