missing translation for 'onlineSavingsMsg'
Learn More

Procymidox-d5, TRC

Product Code. 30422530
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422530 25 mg -
30413657 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30422530 Supplier Toronto Research Chemicals Supplier No. TRCP75585725MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C13H6D5Cl2NO2
Molecular Weight (g/mol) 289.17
Synonym 3-(3,5-Dichlorophenyl)-1,5-dimethyl-3-azabicyclo[3.1.0]hexane-2,4-dione-d5, 1,2-Dimethyl-N-(3,5-dichlorophenyl)cyclopropanedicarboximide-d6, Dicyclidine-d5, Dicyclidine-d5, Kenolex-d5, Procilex-d5, Procymidone-d5, Procymidor-d5, Procymidox-d5, Salithiex-d5, Sialex-d5, Sumilex-d5, Sumilex-d5, Sumisclex-d5
SMILES O=C(N1C2=CC(Cl)=CC(Cl)=C2)C3(C)C([2H])([2H])C3(C([2H])([2H])[2H])C1=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C13H11Cl2NO2/c1-12-6-13(12,2)11(18)16(10(12)17)9-4-7(14)3-8(15)5-9/h3-5H,6H2,1-2H3/i1D3,6D2
Percent Purity >95%
Chemical Name or Material Procymidox-d5
Grade HPLC
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.