Learn More
- Home
- Products
- Flasks
- Flask Accessories
- Corning™ ProCulture™ Fittings
Corning™ ProCulture™ Fittings
Product Code. 10718764 Shop All Corning Products
Change view
Click to view available options
Product Type:
Combination Sidearm
Dual Sidearm
Media Handling Dual Sidearm
Sidearm
Triple Sidearm
Description:
1 Tube
2 Tubes, Angled
2 Tubes, Straight
3 Tubes, Angled
Diameter (Metric) Tubing:
18 mm
3.17 mm
6.35 mm
6.35/3.17 mm
For Use With (Equipment):
1 L Sidearm Spinner Flask
15 L Sidearm Spinner Flask
3 L Sidearm Spinner Flask
500 L Sidearm Spinner Flask
6 L Sidearm Spinner Flask
8 L Sidearm Spinner Flask
8, 15 L Sidearm Spinner Flask
Large Spinner Flasks w/ Vertical Sidearms
Unit Size:
Each
| Product Code. | Product Type | Description | Diameter (Metric) Tubing | For Use With (Equipment) | unitSize |
|---|---|---|---|---|---|
| 10718764 | Dual Sidearm | 2 Tubes, Angled | 3.17 mm | 6 L Sidearm Spinner Flask | Each |
| 10676273 | Dual Sidearm | 2 Tubes, Angled | 3.17 mm | 1 L Sidearm Spinner Flask | Each |
| 10184112 | Dual Sidearm | 2 Tubes, Angled | 6.35/3.17 mm | 500 L Sidearm Spinner Flask | Each |
| 10032212 | Combination Sidearm | 2 Tubes, Angled | 6.35 mm | 3 L Sidearm Spinner Flask | Each |
| 10377142 | Dual Sidearm | 2 Tubes, Angled | 3.17 mm | 3 L Sidearm Spinner Flask | Each |
| 10072102 | Dual Sidearm | 2 Tubes, Angled | 6.35/3.17 mm | 8 L Sidearm Spinner Flask | Each |
| 10032142 | Dual Sidearm | 2 Tubes, Angled | 3.17 mm | 8 L Sidearm Spinner Flask | Each |
| 10511853 | Dual Sidearm | 2 Tubes, Angled | 6.35 mm | 1 L Sidearm Spinner Flask | Each |
| 10052152 | Dual Sidearm | 2 Tubes, Angled | 6.35 mm | 3 L Sidearm Spinner Flask | Each |
| 10307292 | Dual Sidearm | 2 Tubes, Angled | 6.35 mm | 8 L Sidearm Spinner Flask | Each |
| 10521853 | Dual Sidearm | 2 Tubes, Angled | 6.35 mm | 6 L Sidearm Spinner Flask | Each |
| 10377382 | Sidearm | 2 Tubes, Angled | 3.17 mm | 1 L Sidearm Spinner Flask | Each |
| 10799524 | Sidearm | 2 Tubes, Angled | 3.17 mm | 6 L Sidearm Spinner Flask | Each |
| 10511683 | Sidearm | 2 Tubes, Angled | 3.17 mm | 8 L Sidearm Spinner Flask | Each |
| 10767424 | Triple Sidearm | 3 Tubes, Angled | 3.17 mm | 6 L Sidearm Spinner Flask | Each |
| 10072162 | Combination Sidearm | 2 Tubes, Angled | 3.17 mm | 3 L Sidearm Spinner Flask | Each |
| 10757424 | Triple Sidearm | 3 Tubes, Angled | 3.17 mm | 3 L Sidearm Spinner Flask | Each |
| 10770295 | Sidearm | 2 Tubes, Angled | 6.35 mm | 8 L Sidearm Spinner Flask | Each |
| 10133732 | Combination Sidearm | 2 Tubes, Angled | 6.35 mm | 1 L Sidearm Spinner Flask | Each |
| 10285792 | Triple Sidearm | 3 Tubes, Angled | 3.17 mm | 1 L Sidearm Spinner Flask | Each |
| 10482802 | Triple Sidearm | 3 Tubes, Angled | 3.17 mm | 8 L Sidearm Spinner Flask | Each |
| 10012122 | Dual Sidearm | 2 Tubes, Angled | 6.35 mm | 15 L Sidearm Spinner Flask | Each |
| 10777614 | Sidearm | 2 Tubes, Angled | 6.35 mm | 15 L Sidearm Spinner Flask | Each |
| 10550723 | Media Handling Dual Sidearm | 2 Tubes, Straight | 6.35 mm | 8, 15 L Sidearm Spinner Flask | Each |
| 10443382 | Sidearm | 1 Tube | 18 mm | Large Spinner Flasks w/ Vertical Sidearms | Each |
| 10377812 | Dual Sidearm | 2 Tubes, Angled | 6.35/3.17 mm | 1 L Sidearm Spinner Flask | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Dual angled sidearm fittings can be used for aseptically transferring medium into or out of angled sidearm spinner flasks or for sparging the medium with gases.
- Fittings are comprised of a PET insert with a fluorocarbon O-Ring and a polypropylene sealing cap.
- Two 316 stainless steel tubes, which extend to the bottom of the flask, are held in place by modified polyphenylene oxide thermoplastic nuts with integrated ferrules.
- The fittings are completely autoclavable.
Spare parts for sidearm fittings, see Cat. Nos. 1395-32LTC, 1395-45LTC, 1395-LTR, and 1395LTMC
Specifications
Specifications
| Description | 2 Tubes, Angled |
| Diameter (Metric) Tubing | 3.17 mm |
| Material | Stainless Steel |
| For Use With (Application) | Used to sparge the cell culture medium with gases to enhance cell production. |
| For Use With (Equipment) | 6 L Sidearm Spinner Flask |
| Includes | Insert, O-ring, sealing cap |
| No. per Case | Each |
| Product Type | Dual Sidearm |
| Autoclavable | Autoclavable |
| Sterility | Non-sterile |