Description
- Hand-held, cordless homogenizer features a simple push-in, pull-out pestle exchange and is ideal for thorough and faster processing of one-to-many samples
- Optional ProCulture™ Matched Pestles and Tubes assure tight tolerances and thorough homogenization and grinding of materials resulting in optimal efficiency of pellet reconstitution or re-suspension; F65000-0005, F65000-0006, F65000-9006
- Optional ProCulture™ Pestles have a unique ribbed shaft that provides precise finger control of homogenization for shear sensitive samples; also fit most motorized mixer adapters; F65000-0001, F65000-0002, F65000-9001, F65000-9002
Specifications
Specifications
| Product Type | Cordless Homogenizer Unit |
| Voltage | 0/1 V |
| Certifications/Compliance | CE |
| Dnase Free | Yes |
| Includes | Homogenizer with Cordless Motor, AA Batteries (2), and Pestle Adapter |
| RNase Free | Yes |
WARNING: This product can expose you to chemicals which are known to the State of California to cause cancer and birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.