missing translation for 'onlineSavingsMsg'
Learn More

PRMT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17628-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16897564

  • 1415.13 NOK / 20µL
Estimated Shipment: 12-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Type I protein arginine N-methyltransferases (PRMTs), such as PRMT3, catalyze the formation of asymmetric N(G),N(G)-dimethylarginine (ADMA) residues in proteins.
Specifications

Specifications

PRMT3
Polyclonal
Unconjugated
PRMT3
HRMT1L3, PRMT3
Rabbit
Antigen Affinity Chromatography
RUO
10196, 71974, 89820
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O60678, O70467, Q922H1
Prmt3
PRMT3 Fusion Protein Ag11877
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers