Learn More
PRM1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15697-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The PRM1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PRM1. This antibody recognizes human, mouse, rat antigen. The PRM1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
PRM1 | |
Polyclonal | |
Unconjugated | |
20 μL | |
5619 | |
IgG |
Western Blot, ELISA | |
553 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |