missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRKX Recombinant Human Protein
PRKX is a serine/threonine kinase.
Brand: Invitrogen™ PV4045
Additional Details : Weight : 0.00900kg
Description
PRKX is related to the catalytic subunit of the cAMP-dependent protein kinases.
Specifications
NP_005035 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
PRKX, PKX1 | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
68.4kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
PKA Kinase Family | |
Recombinant | |
Baculovirus |