missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PRKCH (Human) Recombinant Protein

Human PRKCH full-length ORF ( AAH01000, 1 a.a. - 41 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00005583-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16129571

  • 4115.00 NOK / 10µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MALEIVDNEMRFYEVYRSTYERLSLWAWDVNRSQKEGKCEE
Specifications

Specifications

AAH01000
protein kinase C, eta
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PRKCH
GST
5583
Wheat germ expression system
10 μg
MGC26269, MGC5363, PKC-L, PKCL, PRKCL, nPKC-eta
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers