missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PRKCBP1 Polyclonal antibody specifically detects PRKCBP1 in Human,Mouse samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications
Specifications
| Antigen | PRKCBP1 |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 610 SE |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ESP-2, HED-2, zyxin, Zyxin-2 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 959-1188 of human PRKCBP1 (NP_898868.1).,, Sequence:, LELTMAEMRQSLEQERDRLIAEVKKQLELEKQQAVDETKKKQWCANCKKEAIFYCCWNTSYCDYPCQQAHWPEHMKSCTQSATAPQQEADAEVNTETLNKSSQGSSSSTQSAPSETASASKEKETSAEKSKESGSTLDLSGSRETPSSILLGSNQGSVSKRCDKQPAYAPTTTDHQPHPNYPAQKYHSRSNKSSWSSSDEKRGSTRSDHNTSTSTKSLLPKESRLDTFWD |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?