Learn More
Epredia™ PrintMate™ Slotted Cassettes
Product Code. p-6221510 Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 1000
Pack of 750
Quantity:
10 x 75 cassettes (750)/cs
4 x 250 cassettes (1000)/cs
Product Code. Color unitSize Quantity
16323996 Beige Pack of 1000 4 x 250 cassettes (1000)/cs
16333996 Blue Pack of 1000 4 x 250 cassettes (1000)/cs
16343996 Green Pack of 1000 4 x 250 cassettes (1000)/cs
16353996 Gray Pack of 1000 4 x 250 cassettes (1000)/cs
16363996 Lavender Pack of 1000 4 x 250 cassettes (1000)/cs
16373996 Light Green Pack of 1000 4 x 250 cassettes (1000)/cs
16383996 Lilac Pack of 1000 4 x 250 cassettes (1000)/cs
16393996 Orange Pack of 1000 4 x 250 cassettes (1000)/cs
16304006 Pink Pack of 1000 4 x 250 cassettes (1000)/cs
16314006 Tan Pack of 1000 4 x 250 cassettes (1000)/cs
16324006 White Pack of 1000 4 x 250 cassettes (1000)/cs
16334006 Yellow Pack of 1000 4 x 250 cassettes (1000)/cs
16374026 Beige Pack of 750 10 x 75 cassettes (750)/cs
16384026 Blue Pack of 750 10 x 75 cassettes (750)/cs
16394026 Green Pack of 750 10 x 75 cassettes (750)/cs
16304036 Gray Pack of 750 10 x 75 cassettes (750)/cs
16314036 Lavender Pack of 750 10 x 75 cassettes (750)/cs
16324036 Light Green Pack of 750 10 x 75 cassettes (750)/cs
16334036 Lilac Pack of 750 10 x 75 cassettes (750)/cs
16344036 Orange Pack of 750 10 x 75 cassettes (750)/cs
16354036 Pink Pack of 750 10 x 75 cassettes (750)/cs
16364036 Tan Pack of 750 10 x 75 cassettes (750)/cs
16374036 White Pack of 750 10 x 75 cassettes (750)/cs
16384036 Yellow Pack of 750 10 x 75 cassettes (750)/cs
24 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16323996
Marke: Epredia™ B851090BE
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. PrintMate Cassettes provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.
Designed for performance and user convenience, PrintMate Slotted Cassettes have been optimized for superior workflow and printing with the PrintMate Classic and PrintMate AS printing systems. The even, textured printing face provides the optimal surface for high-quality printing every time. For additional information, the right and left sides are textured for clear handwriting with suitable pens. The lid catches have been adjusted for ease of use with low opening and closing force, while still giving the security needed while processing.Available in tubes for automatic printing or loose for manual use, and in twelve colors for enhanced workflow identification.
TRUSTED_SUSTAINABILITY
Spezifikation
Pore Style Slots
Closure Included Separate lids included
Quantity 4 x 250 cassettes (1000)/cs
Closure Type Reversible
PrintMate Compatible Yes
Color Beige
Product Type Slotted Cassette