Learn More
Epredia™ PrintMate™ Slotted Cassettes
Product Code. 16383996 Shop All Epredia Products
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Quantity:
10 x 75 cassettes (750)/cs
4 x 250 cassettes (1000)/cs
Unit Size:
Pack of 1000
Pack of 750
Product Code. 16383996
Brand: Epredia™ B851090LI
Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. PrintMate Cassettes provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.
Designed for performance and user convenience, PrintMate Slotted Cassettes have been optimized for superior workflow and printing with the PrintMate Classic and PrintMate AS printing systems. The even, textured printing face provides the optimal surface for high-quality printing every time. For additional information, the right and left sides are textured for clear handwriting with suitable pens. The lid catches have been adjusted for ease of use with low opening and closing force, while still giving the security needed while processing.Available in tubes for automatic printing or loose for manual use, and in twelve colors for enhanced workflow identification.
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Slots
Closure Included Separate lids included
Quantity 4 x 250 cassettes (1000)/cs
Closure Type Reversible
PrintMate Compatible Yes
Color Lilac
Product Type Slotted Cassette