Learn More
Epredia™ PrintMate™ Biopsy Cassettes
Product Code. 16304026 Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Quantity:
10 x 75 cassettes (750)/cs
4 x 250 cassettes (1000)/cs
Unit Size:
Pack of 1000
Pack of 750
Product Code. Color Quantity unitSize
16304026 Light Green 10 x 75 cassettes (750)/cs Pack of 750
16313986 Beige 4 x 250 cassettes (1000)/cs Pack of 1000
16232321 Blue 4 x 250 cassettes (1000)/cs Pack of 1000
16323986 Green 4 x 250 cassettes (1000)/cs Pack of 1000
16333986 Gray 4 x 250 cassettes (1000)/cs Pack of 1000
16343986 Lavender 4 x 250 cassettes (1000)/cs Pack of 1000
16353986 Light Green 4 x 250 cassettes (1000)/cs Pack of 1000
16363986 Lilac 4 x 250 cassettes (1000)/cs Pack of 1000
16373986 Orange 4 x 250 cassettes (1000)/cs Pack of 1000
16383986 Pink 4 x 250 cassettes (1000)/cs Pack of 1000
16393986 Tan 4 x 250 cassettes (1000)/cs Pack of 1000
16303996 White 4 x 250 cassettes (1000)/cs Pack of 1000
16313996 Yellow 4 x 250 cassettes (1000)/cs Pack of 1000
16354016 Beige 10 x 75 cassettes (750)/cs Pack of 750
16364016 Blue 10 x 75 cassettes (750)/cs Pack of 750
16374016 Green 10 x 75 cassettes (750)/cs Pack of 750
16384016 Gray 10 x 75 cassettes (750)/cs Pack of 750
16394016 Lavender 10 x 75 cassettes (750)/cs Pack of 750
16314026 Lilac 10 x 75 cassettes (750)/cs Pack of 750
16324026 Orange 10 x 75 cassettes (750)/cs Pack of 750
16334026 Pink 10 x 75 cassettes (750)/cs Pack of 750
16344026 Tan 10 x 75 cassettes (750)/cs Pack of 750
16354026 White 10 x 75 cassettes (750)/cs Pack of 750
16364026 Yellow 10 x 75 cassettes (750)/cs Pack of 750
24 options
Product Code. 16304026
Brand: Epredia™ B851110LG
Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. PrintMate Cassettes provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.
Designed for performance and user convenience, the PrintMate cassette range has been optimized for superior workflow and printing with the PrintMate Classic and PrintMate AS printing systems. The even, textured printing face provides the optimal surface for high-quality printing every time. For additional information, the right and left sides are textured for clear handwriting with suitable pens. The lid catches have been adjusted for ease of use with low force opening and closing, while still giving the security needed while processing.Available in tubes for automatic loading or loose for manual use, and in twelve colors for enhanced workflow identification.
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Small Square Pores
Closure Included Separate lids included
Quantity 10 x 75 cassettes (750)/cs
Closure Type Reversible
PrintMate Compatible Yes
Color Light Green
Product Type PrintMate Biopsy Cassette for PrintMate Printers