missing translation for 'onlineSavingsMsg'
Learn More

Primulin, MP Biomedicals™

Product Code. p-4405826 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11465382

Brand: MP Biomedicals 0219545405

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 8064-60-6
Molecular Formula C21H14N3NaO3S3
Molecular Weight (g/mol) 475.531
MDL Number MFCD00005798
InChI Key RSRNHSYYBLEMOI-UHFFFAOYSA-M
Synonym Direct Yellow 59, C.I. 49000
PubChem CID 3769888
IUPAC Name sodium;2-[2-(4-aminophenyl)-1,3-benzothiazol-6-yl]-6-methyl-1,3-benzothiazole-7-sulfonate
SMILES CC1=C(C2=C(C=C1)N=C(S2)C3=CC4=C(C=C3)N=C(S4)C5=CC=C(C=C5)N)S(=O)(=O)[O-].[Na+]

Specifications

Formula Weight 475.5
Color Brown/Yellow
Quantity 5 g
Physical Form Powder
Chemical Name or Material Primulin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.