Learn More
Primulin, MP Biomedicals™
Product Code. 11465382 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11465382
Brand: MP Biomedicals 0219545405
Only null left
Chemical Identifiers
CAS 8064-60-6
Molecular Formula C21H14N3NaO3S3
Molecular Weight (g/mol) 475.531
MDL Number MFCD00005798
InChI Key RSRNHSYYBLEMOI-UHFFFAOYSA-M
Synonym Direct Yellow 59, C.I. 49000
PubChem CID 3769888
IUPAC Name sodium;2-[2-(4-aminophenyl)-1,3-benzothiazol-6-yl]-6-methyl-1,3-benzothiazole-7-sulfonate
SMILES CC1=C(C2=C(C=C1)N=C(S2)C3=CC4=C(C=C3)N=C(S4)C5=CC=C(C=C5)N)S(=O)(=O)[O-].[Na+]
Specifications
Formula Weight 475.5
Color Brown/Yellow
Quantity 5 g
Physical Form Powder
Chemical Name or Material Primulin