Learn More
Zeiss™ Primovert Inverted Microscope with Phototube
Description
- The Primovert is perfectly suited to your cell culture laboratory.
- It enables fast, efficient investigations of both unstained cells in phase contrast and GFP-labeled cells in fluorescence contrast.
- It fits straight into your laminar flow cabinet to work directly in a sterile environment.
- The Primovert is easy to use and with little set up the system is ready to use out of the box.
- Automatic light source switch-off (in addition to manual on/off-function): Primo Vert switches off the light automatically after 15 min - saving energy and protecting the stand
- Modular illumination: halogen or long lasting LED
- High quality material: the stand is made of metal and all operating elements are coated with a special touch-friendly rubber compound
- Universal phase slider (optional) for all objectives: for the ease of use there is one phase ring (Ph1) available for 10x, 20x and 40x - this means not having to change the position of the phase slider when changing objectives. In short: time-saving and economical
- Increased working distance thanks to easy removal of the condenser, e.g., for roller-bottles
- Objective indicators for fast identification of magnification
- Integrated carrying handle
- Award-winning design The new Primo Vert inverted microscope is focused on the essentials: excellent optics, high-durability quality materials, ease of use and an appealing design.
- The Primo Vert addresses a wide range of applications: from routine laboratories for live cell inspection to cutting edge research laboratories which require - in addition to research microscopes - compact and reliable routine microscopes for quick and efficient viewing of living cells.
- Typical applications include cancer and HIV research, human, animal and plant genetics, and cell biology in general.
Specifications
Specifications
| Includes | Plan Achromat LD 20 x/0.3 Ph1 and LD 40 x/0.5 Ph1 |
| Objective Types | 4X, 10X, 20X and 40X |
| For Use With (Application) | Cell Culture, Routine Work and Quick and Easy Imaging |