Learn More
Invitrogen™ PRIMER HPLC X 50 NMOL
Product Code. 15490494
Click to view available options
:
50 nmol
Product Code. 15490494
Brand: Invitrogen™ 100032000
PRIMER HPLC X 50 NMOL
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Description HPLC Primer, 50 nmol
Product Type HPLC Primer
Quantity 50 nmol
Grade HPLC