Learn More
Invitrogen™ PrimeFlow™ RNA 100 tests
Product Code. 15940593
Click to view available options
Quantity:
100 tests
Unit Size:
Pack of 100
Product Code. 15940593
Brand: Invitrogen™ CPF210
This is a customizable probe set for use with PrimeFlow Assays.
This is a customizable probe set for use with PrimeFlow Assays.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 660 uL PrimeFlow probe set, store (at -20 C)
Product Line PrimeFlow
Quantity 100 tests