missing translation for 'onlineSavingsMsg'
Learn More

PRIM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11788-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16849093

  • 3828.53 NOK / 150µL
Estimated Shipment: 16-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

PARD proteins, which were first identified in C. elegans, are essential for asymmetric cell division and polarized growth, whereas CDC42 mediates the establishment of cell polarity. The CDC42 GTPase, which is controlled by nucleotide exchange factors and GTPase-activating proteins domain.
Specifications

Specifications

PRIM2
Polyclonal
Unconjugated
PRIM2
DNA primase 58 kDa subunit, DNA primase large subunit, p58, PRIM2, PRIM2A
Rabbit
Antigen Affinity Chromatography
RUO
19076, 301323, 5558
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O89044, P33610, P49643
Prim2
PRIM2 Fusion Protein Ag2370
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers