missing translation for 'onlineSavingsMsg'
Learn More

PRIM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16859093 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16859093 20 μL 20µL
16849093 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16859093

Brand: Proteintech 117881AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PARD proteins, which were first identified in C. elegans, are essential for asymmetric cell division and polarized growth, whereas CDC42 mediates the establishment of cell polarity. The CDC42 GTPase, which is controlled by nucleotide exchange factors and GTPase-activating proteins domain.

Specifications

Antigen PRIM2
Applications Immunocytochemistry, Western Blot, Immunofluorescence
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PRIM2
Gene Accession No. O89044, P33610, P49643
Gene Alias DNA primase 58 kDa subunit, DNA primase large subunit, p58, PRIM2, PRIM2A
Gene Symbols Prim2
Host Species Rabbit
Immunogen PRIM2 Fusion Protein Ag2370
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19076, 301323, 5558
Target Species Mouse, Rat, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.