missing translation for 'onlineSavingsMsg'
Learn More

Prexasertib dihydrochloride, MedChemExpress

Product Code. 16443596
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16443596 25 mg 25mg
16483497 50 mg 50mg
16437478 10 mg 10mg
16429957 5 mg 5mg
16407446 100 mg 100mg
5 options
This item is not returnable. View return policy

Product Code. 16443596

Brand: Medchem Express HY18174A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Prexasertib dihydrochloride (LY2606368 dihydrochloride) is a selective, ATP-competitive second-generation checkpoint kinase 1 (CHK1) inhibitor with a Ki of 0.9 nM and an IC50 of 50=8 nM) and RSK1 (IC50=9 nM). Prexasertib dihydrochloride causes double-stranded DNA breakage and replication catastrophe resulting in apoptosis. Prexasertib dihydrochloride shows potent anti-tumor activity.

Prexasertib dihydrochloride (LY2606368 dihydrochloride) is a selective, ATP-competitive second-generation checkpoint kinase 1 (CHK1) inhibitor with a Ki of 0.9 nM and an IC50 of 50=8 nM) and RSK1 (IC50=9 nM). Prexasertib dihydrochloride causes double-stranded DNA breakage and replication catastrophe resulting in apoptosis. Prexasertib dihydrochloride shows potent anti-tumor activity.

Specifications

Chemical Name or Material Prexasertib dihydrochloride
CAS 1234015-54-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H21Cl2N7O2
Quantity 25 mg
Synonym LY2606368 dihydrochloride
Solubility Information DMSO : 8 mg/mL (18.25 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.28 mM; ultrasonic and warming and heat to 80°C)
SMILES NCCCOC(C=CC=C1OC)=C1C2=CC(NC3=NC=C(C#N)N=C3)=NN2.[H]Cl.[H]Cl
Molecular Weight (g/mol) 438.31
Formula Weight 438.31
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.