Learn More
Thermo Scientific™ Oxoid™ Pressure Gauge for Anaerobic Jar, Oxoid
Description
Pressure Gauge for use with Thermo Scientific Oxoid 3.5L Anaerobic Jar (HP0011A).
Specifications
Specifications
| Type | Pressure Gauge |
| For Use With (Equipment) | Anaerobic Jar |