Learn More
Cole-Parmer™ Stuart™ Pressmatic PTFE Ceramic Dispenser
Product Code. 12837933
Change view
Click to view available options
Volume (Metric):
2 to 10 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
12837933 2 to 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12837933 Supplier Cole-Parmer™ Supplier No. 1011278
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Accuracy <0.7%
Product Type Bottletop Dispenser
Material PTFE, Ceramic
Volume (Metric) 2 to 10 mL
For Use With (Equipment) Bottle
Increments 0.25 mL