missing translation for 'onlineSavingsMsg'
Learn More

Presatovir, MedChemExpress

Product Code. 16457156
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16457156 100 mg 100mg
16499687 5 mg 5mg
16497158 10 mg 10mg
16463187 50 mg 50mg
4 options
This item is not returnable. View return policy

Product Code. 16457156

Brand: Medchem Express HY16727

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Presatovir (GS-5806) is an orally bioavailable RSV fusion inhibitor with a mean EC50 value of 0.43 nM.

Presatovir (GS-5806) is an orally bioavailable RSV fusion inhibitor with a mean EC50 value of 0.43 nM.

Specifications

Chemical Name or Material Presatovir
CAS 1353625-73-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H30ClN7O3S
Quantity 100 mg
Synonym GS-5806
Solubility Information DMSO : 6 mg/mL (11.28 mM; Need ultrasonic and warming) ∣H2O : ≥ 0.1 mg/mL (0.19 mM)
SMILES O=C(C(C=C(Cl)C=C1)=C1NS(C)(=O)=O)N(CCCC2)[C@@H]2C3=NN4C(N=C(N5CC[C@H](N)C5)C(C)=C4)=C3
Molecular Weight (g/mol) 532.06
Formula Weight 532.06
Percent Purity 99.94%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.