missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Preprotachykinin-1 Antibody [Allophycocyanin], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
Preprotachykinin-1 Polyclonal antibody specifically detects Preprotachykinin-1 in Human samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | Preprotachykinin-1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | APC |
| Formulation | PBS |
| Gene Alias | Hs.2563, NK2, NKNA, NPK, PPT, protachykinin-1, TAC1, TAC2 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-129 of human Preprotachykinin-1 (NP_003173.1).,, Sequence:, MKILVALAVFFLVSTQLFAEEIGANDDLNYWSDWYDSDQIKEELPEPFEHLLQRIARRPKPQQFFGLMGKRDADSSIEKQVALLKALYGHGQISHKRHKTDSFVGLMGKRALNSVAYERSAMQNYERRR |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?