Learn More
Kappes Systeme™ Premium Storage Bins
Specifications
Specifications
| Type | Storage Bin |
| Material | Polyethylene |
| Color | Red |
| Shape | Rectangular |
| For Use With | Feed and Roller Conveyor |
| For Use With (Application) | Food Storage |
| Style | Size 4 |
| Length (Metric) | 350 mm |
| Width (Metric) | 200 mm |
| Height (Metric) | 150 mm |