Learn More
Kappes Systeme™ Premium Storage Bins
Product Code. 12602065
Encompass_Preferred
Change view
Click to view available options
Color:
Blue
Gray
Green
Red
Yellow
Style:
Size 1
Size 2
Size 3
Size 4
Unit Size:
Each
Product Code. Color Style unitSize
12602065 Yellow Size 3 Each
12652045 Blue Size 1 Each
12672045 Yellow Size 1 Each
12642045 Green Size 1 Each
12632055 Blue Size 2 Each
12642055 Red Size 2 Each
12672055 Blue Size 3 Each
12692055 Red Size 3 Each
12612065 Gray Size 2 Each
12682055 Green Size 3 Each
12651775 Red Size 4 Each
12611775 Blue Size 4 Each
12621775 Yellow Size 4 Each
12601775 Green Size 4 Each
12641775 Gray Size 4 Each
15 options
Product Code. 12602065
Brand: Kappes Systeme™ 6002.00.0252
This item requires a price update prior to being ordered.
Request a Price for this item here.
Specifications
Type Storage Bin
Material Polyethylene
Color Yellow
Shape Rectangular
For Use With Feed and Roller Conveyor
For Use With (Application) Food Storage
Style Size 3
Length (Metric) 350 mm
Width (Metric) 200 mm
Height (Metric) 200 mm