Learn More
Tocris Bioscience™ Pregnenolone sulfate sodium salt
GREENER_CHOICE
NMDA potentiator; also GABAA inhibitor and TRPM3 activator
Brand:  Tocris Bioscience™ 5376/50
Encompass
Product Code. 15737356
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Pregnenolone sulfate sodium salt is a highly potent neurosteroid with multiple actions. Enhances intracellular Ca2+ upon NMDA receptor-mediated synaptic activity (EC50 ∽ 2 pM) and potentiates NMDA responses. Inhibitor at GABAA receptors (IC50 values are 0.4 to >300 μM, depending on specific subunit conformation). Activates CREB phosphorylation, augments LTP and cognitive behavior in rat memory models. Also activates TRPM3 channels.